Learn More
American Research Products Inc Rabbit plasmin antiplasmin com
Supplier:  American Research Products Inc CSBE08784RB
Encompass
Rabbit plasmin-antiplasmin compleX PAP ELISA Kit from CUSABIO Detection Range 0 25 ng/mL-16 ng/mL Sensitivity 0 06 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29208