Learn More
Cayman Chemical Rabeprazole sulfon 10mg
SDP
Supplier:  Cayman Chemical 3507610 MG
Encompass_Preferred
A metabolite of rabeprazole; has been found as an impurity in bulk preparations of rabeprazole
Catalog No. 50-291-1434