Learn More
Fujirebio Diagnostics Inc FITC Anti-Rabies Monoclonal Globulin
Supplier:  Fujirebio Diagnostics Inc 800092
Encompass
The FDI FITC ANTI-RABIES MONOCLONAL GLOBULIN is used in the direct fluorescent antibody procedure for the in vitro detection of rabies in brains and submaxillary glands.
Catalog No. NC9554844