Learn More
ABclonal Technology RABIF Rabbit pAb
SDP
Supplier:  ABclonal Technology A150901000UL
Encompass_Preferred
This gene encodes a member of the SCE4/YPT1/RAB family of small GTP-binding proteins that are involved in the regulation of intracellular vesicular transport. This protein stimulates GTP-GDP exchange in SEC4, and to a lesser extent in YPT1 and RAB3A, and may play a general role in vesicular transport.
Catalog No. 50-001-05393