Learn More
ABclonal Technology RABL3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143271000UL
Encompass_Preferred
Predicted to enable GTP binding activity, GTPase activity, and protein homodimerization activity. Involved in regulation of Ras protein signal transduction and regulation of protein lipidation. Predicted to be active in endomembrane system. Implicated in pancreatic cancer.
Catalog No. 50-001-09024