Learn More
Medchemexpress LLC HY-14720 10mg Medchemexpress, Rabusertib CAS:911222-45-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1472010MG
Encompass_Preferred
Medchemexpress, HY-14720 10mg Rabusertib CAS:911222-45-2 Rabusertib (LY2603618) is a potent and selective inhibitor of Chk1 with an IC50 of 7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9060
 Restricted Item