Learn More
Medchemexpress LLC Rac 1,2-bis-palmitoyl-3-chloropropanediol-d5 | 1185057-55-9 | MFCD09840072 | 97.6% | C35H62D5ClO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY143699S10MG
Encompass_Preferred
(Rac)-1,2-Bis-palmitoyl-3-chloropropanediol-d5 is the deuterium-labeled analogue of 1,2-bis-palmitoyl-3-chloropropanediol, provided as a white to off-white solid. It serves as a tracer and internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. Recommended storage is sealed and protected from moisture at -20°C, with solutions stored at -80°C for extended stability.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Suitable as a tracer for quantitative studies.
  • High purity (97.6%).
  • Solid form, white to off-white color.
  • Molecular formula C35H62D5ClO4 and molecular weight 592.39.
  • Available in small-mass quantities suitable for analytical workflows.
Catalog No. 50-002-58547