Learn More
Medchemexpress LLC Rac-1,2-dioleoyl-3-chloropropanediol-d5 | 1246833-00-0 | >99.0% | 644.46 | C39H66D5ClO4 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HYW587715S2.5MG
Encompass_Preferred
(Rac)-1,2-Dioleoyl-3-chloropropanediol-d5 is a deuterium-labeled lipid reagent for laboratory research, used primarily as a tracer or an internal standard in quantitative analyses. It is suited for isotope-labeling studies and for improving accuracy and precision in mass spectrometry and NMR assays.
  • Deuterium-labeled (d5) analog for isotope tracing and quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS assays.
  • High chemical purity (>99.0%) for reliable analytical results.
  • Molecular weight 644.46 and formula C39H66D5ClO4 for accurate mass calculations.
  • Provided in a small premeasured quantity to support analytical workflows.
Catalog No. 50-002-83957