Learn More
Medchemexpress LLC Rac-3-hydroxyphenylglycine | 31932-87-3 | MFCD00673854 | 99.6% | 167.16 | C8H9NO3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100841100MG
Encompass_Preferred
(Rac)-3-Hydroxyphenylglycine is a racemic mixture of the S- and R-enantiomers of 3-hydroxyphenylglycine supplied as a powder for research use. The S-enantiomer is a reported potent and selective agonist of the metabotropic glutamate receptor 1 (mGluR1). The substance has formula C8H9NO3, molecular weight 167.16, and is provided with analytical purity and storage recommendations.
Catalog No. 50-002-11725