Learn More
Medchemexpress LLC Rac-3-hydroxyphenylglycine | 31932-87-3 | MFCD00673854 | 99.6% | 167.16 | C8H9NO3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10084125MG
Encompass_Preferred
(Rac)-3-Hydroxyphenylglycine is the racemic mixture of (S)- and (R)-3-hydroxyphenylglycine supplied as a research reagent and characterized as an mGluR1 agonist. The compound is provided with analytical data and documentation for laboratory use and is intended for in vitro pharmacology and neuroscience studies. Not for human or clinical use.
  • High purity suitable for research applications.
  • Molecular weight 167.16 and formula C8H9NO3 for unambiguous identification.
  • Available in small milligram pack sizes for screening and assay development.
  • Supplied with a product datasheet containing CAS and analytical information.
Catalog No. 50-002-12339