Learn More
Medchemexpress LLC (Rac)-5-Hydroxymethyl Tolterodine | 200801-70-3 | 99.1% | C22H31NO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY76570500MG
Encompass_Preferred
(Rac)-5-Hydroxymethyl Tolterodine, also known as (Rac)-Desfesoterodine, is an active metabolite of Tolterodine. It acts as a mAChR antagonist, targeting M1-M5 receptors with specific Ki values ranging from 2.3 nM to 2.9 nM. This compound is primarily utilized in research related to overactive bladder. It presents as a solid, white to off-white substance.
  • Functions as a mAChR antagonist
  • Targets M1, M2, M3, M4, and M5 receptors
  • Used for overactive bladder research
  • Solid, white to off-white appearance
  • Molecular weight of 341.50
  • Inhibits carbachol-induced contraction in guinea-pig urinary bladder strips
  • Exhibits significant muscarinic receptor binding activity
Catalog No. 50-004-34022