Learn More
Medchemexpress LLC (Rac)-5-Hydroxymethyl Tolterodine hydrochloride | 250214-40-5 | C22H32ClNO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY76570A500MG
Encompass_Preferred
(Rac)-5-Hydroxymethyl Tolterodine hydrochloride is an active metabolite of Tolterodine, functioning as a muscarinic acetylcholine receptor (mAChR) antagonist. It exhibits Ki values of 2.3 nM, 2 nM, 2.5 nM, 2.8 nM, and 2.9 nM for M1, M2, M3, M4, and M5 receptors, respectively. This compound is suitable for research related to overactive bladder, demonstrating competitive inhibition of carbachol-induced contraction in guinea-pig urinary bladder strips and significant binding to muscarinic receptors.
  • Acts as a muscarinic acetylcholine receptor (mAChR) antagonist
  • Active metabolite of Tolterodine
  • Inhibits carbachol-induced contraction of isolated guinea-pig urinary bladder strips
  • Exhibits significant binding activity to muscarinic receptors in various tissues, with a longer duration in the bladder
  • Suitable for overactive bladder research
Catalog No. 50-004-33661