Learn More
Medchemexpress LLC HY-124540B 5mg Medchemexpress, (Rac)-ABT-202 (dihydrochloride) CAS:1258641-38-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124540B5MG
Encompass_Preferred
Medchemexpress, HY-124540B 5mg (Rac)-ABT-202 (diHCl) CAS:1258641-38-1 (Rac)-ABT-202 dihydrochloride is a racemate of ABT-202. ABT-202 is an agonistor of nicotinic acetylcholine receptors (nAChRs) and can be used as an analgesic. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9667