Learn More
Medchemexpress LLC (Rac)-Acolbifene | 151533-34-5 | 98.2% | 457.56 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16023B100MG
Encompass_Preferred
(Rac)-Acolbifene is the racemic form of EM652, an estrogen receptor antagonist. It exhibits both anti-estrogenic and estrogenic activities. This compound features a piperidine ring and has a good pharmacological profile, with a relative binding affinity (RBA) of 380.
  • It is an estrogen receptor antagonist.
  • It demonstrates anti-estrogenic and estrogenic activities.
  • It contains a piperidine ring.
  • It shows a good pharmacological profile.
  • It has a relative binding affinity (RBA) of 380.
  • It shows an inhibitory effect in T-47D cells with an IC50 value of 0.110 nM.
  • In vivo administration shows significant antiuterotrophic inhibitions in ovariectomized mice.
Catalog No. 50-003-93205