Learn More
Medchemexpress LLC (Rac)-Acolbifene | 151533-34-5 | 98.2% | 457.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16023B
Encompass_Preferred
(Rac)-Acolbifene is the racemic form of EM652, an estrogen receptor antagonist, which exhibits both anti-estrogenic and estrogenic activities. It contains a piperidine ring and demonstrates a good pharmacological profile with a relative binding affinity (RBA) of 380.
  • Has anti-estrogenic and estrogenic activities.
  • Contains a piperidine ring.
  • Shows good pharmacological profile, with a relative binding affinity (RBA) of 380.
  • For research use only.
Catalog No. 50-003-93007