Learn More
Medchemexpress LLC (Rac)-Acolbifene (EM-343; (Rac)-EM-652) | 151533-34-5 | 98.2% | 457.56 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16023B10MM/1ML
Encompass_Preferred
(Rac)-Acolbifene (EM-343; (Rac)-EM-652) is the racemic form of EM652, an estrogen receptor antagonist, exhibiting both anti-estrogenic and estrogenic activities. This compound contains a piperidine ring and demonstrates a good pharmacological profile with a relative binding affinity (RBA) of 380. It shows an inhibitory effect in T-47D cells and antiuterotrophic inhibitions in ovariectomized mice.
  • Racemic form of EM652.
  • Acts as an estrogen receptor antagonist.
  • Exhibits anti-estrogenic and estrogenic activities.
  • Contains a piperidine ring.
  • Demonstrates a good pharmacological profile with a relative binding affinity (RBA) of 380.
  • Inhibits T-47D cell growth with an IC50 value of 0.110 nM.
  • Induces antiuterotrophic inhibitions in ovariectomized mice.
Catalog No. 50-003-93124