Learn More
Medchemexpress LLC (Rac)-AMG8379 | 1641574-26-6 | 99.3% | 543.93 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY108425B1MG
Encompass_Preferred
(Rac)-AMG8379 ((Rac)-AMG8380) is a racemate of AMG8379. AMG8379 is a potent, orally active, and selective sulfonamide antagonist of NaV1.7, with IC50s of 8.5 and 18.6 nM for hNaV1.7 and mNaV1.7, respectively. It is suitable for laboratory centrifugation and storage.
  • Potent, orally active, and selective sulfonamide antagonist of NaV1.7
  • IC50s of 8.5 nM for hNaV1.7
  • IC50s of 18.6 nM for mNaV1.7
Catalog No. 50-003-47150