Learn More
Medchemexpress LLC (Rac)-AMG8379 | 1641574-26-6 | 99.3% | 543.93 g/mol | C25H16ClF2N3O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY108425B5MG
Encompass_Preferred
(Rac)-AMG8379 is the racemic form of AMG8379, a potent, orally active sulfonamide antagonist of the NaV1.7 voltage-gated sodium channel. Supplied as a solid powder for research use, it exhibits low-nanomolar potency and is used to study NaV1.7-mediated pain pathways and sodium-channel pharmacology.
  • Racemic sulfonamide NaV1.7 antagonist.
  • Low-nanomolar potency (hNaV1.7 IC50 8.5 nM; mNaV1.7 IC50 18.6 nM).
  • High purity (99.25%).
  • Solid powder form, suitable for storage and formulation.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
Catalog No. 50-002-63709