Learn More
Medchemexpress LLC Rac-anemonin | 90921-11-2 | 98.2% | 192.17 | C10H8O4 | 10 MG

Supplier: Medchemexpress LLC HYN0278A10MG
(Rac)-Anemonin is a racemic natural-product-derived small molecule used in inflammation-related research. It is reported as an iNOS and PKC-θ dual inhibitor and is supplied with high purity for biochemical and cell-based studies.
- High purity 98.2% suitable for analytical and in vitro assays.
- Molecular weight 192.17 g/mol and formula C10H8O4.
- CAS number 90921-11-2 for unambiguous identification.
- Available as a 10 mg pack for small-scale experiments.
- Suitable for studies of inflammation-related pathways and targets such as iNOS and PKC-θ.
- For research use only; not intended for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.