Learn More
Medchemexpress LLC (rac)-AR-13503 | 1254032-16-0 | 99.1% | 321.37 | C19H19N3O2 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12563950MG
Encompass_Preferred
(rac)-AR-13503 is the racemic active metabolite of AR-13324 (netarsudil) that functions as a Rho-associated kinase (ROCK) and protein kinase C (PKC) inhibitor for preclinical research. It is used to study angiogenesis, retinal pigment epithelium barrier function, and ocular disease models, and is supplied as a high-purity reagent suitable for in vitro and in vivo experiments.
  • Rho-associated kinase and protein kinase C inhibitor.
  • Used in studies of angiogenesis and retinal disease models.
  • High purity for reproducible experimental results.
  • Available in milligram quantities for in vitro and in vivo work.
  • Chemical formula C19H19N3O2 and molecular weight 321.37.
Catalog No. 50-002-37867