Learn More
Medchemexpress LLC rac-AR-13503 (rac-AR-13324 M1 metabolite) | 1254032-16-0 | 99.1% | 321.37 g/mol | C19H19N3O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12563910MG
Encompass_Preferred
(rac)-AR-13503 is a racemic small-molecule metabolite of an investigational ROCK/PKC inhibitor used in preclinical ocular research. It inhibits angiogenesis and increases retinal pigment epithelium permeability, making it useful for studies of neovascularization and retinal barrier function. Supplied for laboratory research use only.
Catalog No. 50-002-37202