Learn More
Medchemexpress LLC (rac)-AR-13503 (rac-AR-13324 M1 metabolite) | 1254032-16-0 | 99.1% | 321.37 g/mol | C19H19N3O2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12563925MG
Encompass_Preferred
(rac)-AR-13503 (CAS 1254032-16-0) is a research small molecule described as the M1 metabolite of netarsudil. It functions as a ROCK and PKC inhibitor and has been reported to inhibit angiogenesis, increase retinal pigment epithelium permeability, and reduce aberrant neovascularization in oxygen-induced retinopathy models. Supplied in milligram-scale quantities for in vitro and in vivo research use.
  • Inhibits Rho-associated kinases and protein kinase C.
  • Demonstrated anti-angiogenic activity in retinal models.
  • Enhances retinal pigment epithelium permeability.
  • Suitable for pharmacology and ophthalmic research applications.
  • Provided in milligram-scale packaging for research experiments.
Catalog No. 50-002-37866