Learn More
Medchemexpress LLC (Rac)-Bepotastine (besilate) | 1415692-17-9 | 99.5% | C27H31CIN2O6S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYA0015A
Encompass_Preferred
(Rac)-Bepotastine (besilate) is an isomer of Bepotastine (besilate) and serves as an experimental control. Bepotastine besilate is a selective, orally active, second-generation histamine H1 receptor antagonist. It can suppress the expression of nerve growth factor (NGF) and has potential for research related to allergic rhinitis, allergic conjunctivitis, and urticaria/pruritus.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Stable under recommended storage conditions
  • Incompatible with strong acids/alkalis and strong oxidising/reducing agents
Catalog No. 50-003-58401