Learn More
Medchemexpress LLC (Rac)-Bepotastine besilate | 1415692-17-9 | 99.5% | C27H31ClN2O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA0015A5MG
Encompass_Preferred
(Rac)-Bepotastine besilate is an isomer of Bepotastine (besilate) that serves as an experimental control. This selective, orally active second-generation histamine H1 receptor antagonist can suppress nerve growth factor (NGF) expression. It has potential for research related to allergic rhinitis, allergic conjunctivitis, and urticaria/pruritus.
  • Selective, orally active second-generation histamine H1 receptor antagonist
  • Suppresses nerve growth factor (NGF) expression
  • Potential research applications in allergic rhinitis, allergic conjunctivitis, and urticaria/pruritus
  • Used as an experimental control for Bepotastine (besilate)
Catalog No. 50-004-33307