Learn More
Strem, An Ascensus Company rac-BI-DIME min. 97% 1g
SDP
Supplier:  Strem, An Ascensus Company 1562051G
Encompass
CAS# 1246888-90-3. 1g. 3-(t-Butyl)-4-(2,6-dimethoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole, min. 97% rac-BI-DIME. . Molecular Weight: 330.36. Molecular Formula: C19H23O3P. Color/form: white xtl. Strem# 15-6205. http://www.strem.com/catalog/v/15-6205/
Catalog No. 50-203-4065