Learn More
Medchemexpress LLC Rac-BL-918 | 2435589-07-2 | 98.9% | 533.44 | C23H15F8N3OS | 100MG

Supplier: Medchemexpress LLC HY124729A100MG
(Rac)-BL-918 is the racemic form of BL-918, a small-molecule activator of UNC-51-like kinase 1 (ULK1) that promotes cytoprotective autophagy. It is intended for preclinical and in vitro research applications where modulation of autophagy or ULK1 activity is required; handle and store according to laboratory safety practices.
- Racemic small-molecule activator of ULK1.
- Induces cytoprotective autophagy in cellular models.
- High purity: 98.9%.
- Molecular weight 533.44 g/mol; formula C23H15F8N3OS.
- Appearance: solid powder; typical package size 100 mg.
- Storage: powder -20°C for long-term; follow solvent storage recommendations when prepared in solution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.