Learn More
Medchemexpress LLC Rac-BL-918 | 2435589-07-2 | 98.9% | 533.44 | C23H15F8N3OS | 10MG
SDP
Supplier:  Medchemexpress LLC HY124729A10MG
Encompass_Preferred
(Rac)-BL-918 is the racemic form of BL-918, a small-molecule activator of UNC-51-like kinase 1 (ULK1) that induces cytoprotective autophagy. It is provided as a powder for research use and is intended for studies of autophagy pathways and neurodegenerative disease models such as Parkinson's disease. Molecular formula C23H15F8N3OS; molecular weight 533.44; CAS 2435589-07-2.
  • Activates ULK1 to induce cytoprotective autophagy.
  • Racemic mixture suitable for research applications.
  • Offered as a powder for convenient handling and storage.
  • High reported purity (98.9%).
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C.
  • Available in small research scales (for example, 1 mg, 5 mg, 10 mg).
Catalog No. 50-002-36719