Learn More
Medchemexpress LLC (Rac)-BL-918 | 2435589-07-2 | 98.9% | 533.44 | C23H15F8N3OS | 10MM 1ML

Supplier: Medchemexpress LLC HY124729A10MM 1ML
(Rac)-BL-918 is the racemic form of BL-918, a small-molecule activator of UNC-51-like kinase 1 (ULK1) that induces cytoprotective autophagy. Provided for research use, it is available as a solid or as a ready-to-use 10 mM solution in DMSO (1 mL) for cellular and preclinical studies probing autophagy and neurodegeneration pathways.
- Molecular weight 533.44 g/mol
- Chemical formula C23H15F8N3OS
- CAS number 2435589-07-2
- Purity 98.9%
- Available as 10 mM solution in DMSO (1 mL) and in solid quantities
- Activates ULK1 and induces autophagy in cellular assays
- Intended for in vitro and preclinical research use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.