Learn More
Medchemexpress LLC HY-124729A 5mg Medchemexpress, (Rac)-BL-918 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124729A5MG
Encompass_Preferred
Medchemexpress, HY-124729A 5mg (Rac)-BL-918 CAS: (Rac)-BL-918 is the racemate of BL-918. BL-918 is a potent activator of UNC-51-like kinase 1 (ULK1) , inducing cytoprotective autophagy for Parkinson’s disease treatment [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2595