Learn More
Medchemexpress LLC Rac-brassinazole | 224047-41-0 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12116110MM 1ML
Encompass_Preferred
(Rac)-Brassinazole is a triazole-type brassinosteroid biosynthesis inhibitor (CAS 224047-41-0) used in plant biology research. Supplied as a 10 mM solution in DMSO (1 ML) for immediate use, it is also available as solid quantities for custom preparation.
  • Triazole-type brassinosteroid biosynthesis inhibitor.
  • Increases inhibition of CYP90B in brassinosteroid biosynthesis.
  • Offered as a ready-to-use 10 mM solution in DMSO (1 ML) and as solids (5 MG-100 MG).
  • Molecular formula C18H18ClN3O; molecular weight 327.81.
  • Supplied with safety data sheet and product data sheet for handling and use.
Catalog No. 50-002-33462