Learn More
Medchemexpress LLC (Rac)-Byakangelicin | 19573-01-4 | 99.92% | 334.32 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN007510MM/1ML
Encompass_Preferred
(Rac)-Byakangelicin is a racemate of Byakangelicin, primarily isolated from the genus Angelica. It functions as an aldose-reductase inhibitor.
  • Acts as an aldose-reductase inhibitor
  • Has an IC₅₀ value of 6.2 μM
  • Primarily isolated from the genus Angelica
Catalog No. 50-004-27895