Learn More
Medchemexpress LLC Rac cis-moxifloxacin-d4 hydrochloride | 1217802-65-7 | 99.78% | C21H21D4ClFN3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY66011S1MG
Encompass_Preferred
Rac cis-Moxifloxacin-d4 (hydrochloride) is the deuterium labeled Moxifloxacin hydrochloride. Moxifloxacin Hydrochloride (BAY 12-8039) is an oral 8-methoxyquinolone antimicrobial for use in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-33826