Learn More
Medchemexpress LLC (Rac)-Cotinine-d3 | 110952-70-0 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1178S
Encompass_Preferred
(Rac)-Cotinine-d3 is the deuterium labeled (Rac)-Cotinine. For research use only. We do not sell to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14449