Learn More
Medchemexpress LLC Rac cotinine-d4 | 350818-68-7 | 99.6% | 180.24 | C10H8D4N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1178S110MG
Encompass_Preferred
(Rac)-Cotinine-d4 is the deuterium-labeled racemic form of cotinine intended for use as an analytical tracer and internal standard in quantitative assays (LC-MS, GC-MS, and NMR). It is supplied in small milligram quantities for research and method development.
  • Deuterium-labeled racemic cotinine (d4) for use as an internal standard.
  • Suitable for LC-MS, GC-MS, and NMR quantitation.
  • High purity (~99.6%) to minimize interference in analytical assays.
  • Provided in small, ready-to-use milligram quantities for analytical workflows.
  • Stable when stored under recommended temperature conditions for reliable performance.
Catalog No. 50-002-96758