Learn More
Medchemexpress LLC (Rac)-CP-601927 hydrochloride | 230615-01-7 | 99.72% | 263.69 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY138879A10MM/1ML
Encompass_Preferred
(Rac)-CP-601927 hydrochloride is the racemate of CP-601927, an agonist for nAChR, with reported Ki values of 1.2 nM for α4β2 nAChR and 102 nM for α3β4 nAChR. It is intended for research use only.
  • Racemic form of CP-601927
  • Acts as a nAChR agonist
  • Exhibits antidepressant-like effects in mice
  • Available as an off-white to light yellow solid
  • Soluble in DMSO and water
Catalog No. 50-003-49705