Learn More
Medchemexpress LLC Rac-dehydrocostus lactone | 74299-48-2 | MFCD00210277 | 99.9% | 230.30 g/mol | C15H18O2 | 10 MG

Supplier: Medchemexpress LLC HYW39162510MG
(Rac)-Dehydrocostus lactone is a sesquiterpene lactone research chemical used in biochemical and pharmacological studies for its reported anti-inflammatory, anticancer, antiviral, and antimicrobial activities. It is supplied as a white to off-white solid at high purity and intended for in vitro research under appropriate laboratory controls.
- High purity: 99.9% (as supplied).
- Molecular formula: C15H18O2; molecular weight: 230.30 g/mol.
- Appearance: white to off-white solid suitable for sample preparation.
- Storage recommendations: powder long-term storage at -20°C; in solvent at -80°C for extended stability.
- Common applications: biochemical assays, cell-based studies, and mechanistic pharmacology research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.