Learn More
Medchemexpress LLC Rac-E1R 100mg | 787623-60-3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY116463D100MG
Encompass_Preferred
(Rac)-E1R is the racemate of E1R, a sigma-1 receptor positive allosteric modulator used for research into cognition and memory disorders. It is provided as a high-purity research compound with supporting SDS and batch documentation.
  • Racemate of E1R, a sigma-1 receptor positive allosteric modulator.
  • Intended for research use only; not for human or veterinary use.
  • Reported purity about 98.48%.
  • Chemical formula C13H16N2O2 and molecular weight 232.28 g/mol.
  • Available in multiple pack sizes, including 100 mg.
  • Supplied with SDS and data sheet for traceability.
Catalog No. 50-002-27133