Learn More
Medchemexpress LLC Rac-E1R 10mM 1mL | 787623-60-3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY116463D10MM 1ML
Encompass_Preferred
(Rac)-E1R is the racemic form of E1R and functions as a sigma-1 receptor positive allosteric modulator (Sig1R PAM) used in preclinical research on cognition and memory disorders. It is supplied in research-grade formulations including a 10 mM solution in DMSO (1 mL) and multiple solid pack sizes. Not for human use.
  • Racemic form of E1R with activity at sigma-1 receptor as a positive allosteric modulator.
  • Available as a 10 mM solution in DMSO for ready-to-use in vitro studies.
  • Also supplied as solid powder in multiple milligram pack sizes for assay preparation.
  • High chemical purity suitable for research applications.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-003-08050