Learn More
Medchemexpress LLC (Rac)-EBET-1055 | 3031540-65-2 | 97.2% | 923.02 g/mol | C51H54N8O9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY161346A10MG
Encompass_Preferred
(Rac)-EBET-1055 is the racemic form of EBET-1055, a bromodomain and extra-terminal (BET) PROTAC degrader supplied for research use. It is provided as a solid suitable for in vitro and in vivo studies of BET protein degradation and epigenetic modulation; the manufacturer supplies solubility and storage recommendations for experimental handling.
  • Racemic BET PROTAC degrader for epigenetic research.
  • Provided in small-mass packages suitable for screening and dosing.
  • High solubility in DMSO (up to 100 mg/mL with ultrasonic assistance).
  • In vivo formulation compatible with PEG300/Tween-80/saline mixtures at ≥2.5 mg/mL.
  • Store at -20 °C, protect from light, and keep under inert atmosphere when recommended.
Catalog No. 50-002-88854