Learn More
Medchemexpress LLC Rac-epoxiconazole | 135319-73-2 | >98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY119683A1MG
Encompass_Preferred
Rac-epoxiconazole is a chiral triazole fungicide supplied as the racemic mixture for research use. It inhibits fungal sterol demethylation, disrupting ergosterol biosynthesis and is used in biochemical and agricultural research to study antifungal activity and mode of action.
  • Chiral triazole fungicide supplied as a racemate.
  • Inhibits fungal sterol demethylation (ergosterol biosynthesis).
  • Broad-spectrum activity against multiple fungal classes.
  • Molecular weight 329.76 and formula C17H13ClFN3O.
  • Supplied in small pack sizes suitable for research (typical 1 mg).
Catalog No. 50-002-33102