Learn More
Medchemexpress LLC (Rac)-Etavopivat | 2622070-93-1 | 99.9% | 457.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139573A100MG
Encompass_Preferred
This product is an isomer of Etavopivat. Etavopivat is an orally active erythrocyte pyruvate kinase-R activator that can be used in studies of sickle cell disease and other haemoglobinopathies. For research use only.
  • Improves haemoglobin-oxygen affinity and reduces the sickle point in human red blood cells.
  • Causes an increase in 2,3-DPG and ATP in crab-eating monkeys.
Catalog No. 50-003-49776