Learn More
Medchemexpress LLC (Rac)-Etavopivat | 2622070-93-1 | 99.9% | 457.50 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139573A
Encompass_Preferred
(Rac)-Etavopivat, also known as (Rac)-FT-4202, is an isomer of Etavopivat. This compound acts as an orally active erythrocyte pyruvate kinase-R (PKR) activator, making it suitable for studies related to sickle cell disease and other hemoglobinopathies.
  • Isomer of Etavopivat
  • Orally active erythrocyte pyruvate kinase-R (PKR) activator
  • Solid appearance, white to off-white color
  • Can be used in studies of sickle cell disease and other hemoglobinopathies
  • Improves hemoglobin-oxygen affinity and reduces the sickle point (PoS) in human red blood cells (In Vitro)
  • Causes an increase in 2,3-DPG and ATP in crab-eating monkeys at doses of 8 mg/kg and 22 mg/kg (In Vivo)
Catalog No. 50-003-49953