Learn More
Medchemexpress LLC (Rac)-etavopivat | 2622070-93-1 | MFCD34578328 | 99.9% | 457.50 | C22H23N3O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY139573A5MG
Encompass_Preferred
(Rac)-Etavopivat is the racemic isomer of etavopivat, an orally active erythrocyte pyruvate kinase-R (PKR) activator used in preclinical research on sickle cell disease and other hemoglobinopathies. Supplied as a high-purity research chemical, it is intended for biochemical and cellular studies investigating PKR activation and related mechanisms.
  • Racemic isomer suitable for comparative pharmacology studies.
  • Orally active PKR activator mechanism relevant to sickle cell research.
  • High purity 99.9% for reproducible experimental results.
  • Molecular formula C22H23N3O6S and molecular weight 457.50 for accurate calculations.
  • Available in small-vial quantities for laboratory use.
  • Stable when stored as recommended, supporting long-term storage.
Catalog No. 50-002-23024