Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-etavopivat | 2622070-93-1 | MFCD34676676 | 99.9% | C22H23N3O6S | 10MG
SDP

Supplier:  Medchemexpress LLC HY139573A10MG

Encompass_Preferred

(Rac)-Etavopivat is the racemic form of etavopivat (FT-4202), an investigational erythrocyte pyruvate kinase (PKR) activator supplied as a solid research chemical for in vitro and preclinical studies. The compound has defined purity, solubility, and storage profiles to support reproducible laboratory experiments.

  • Listed purity 99.87% for analytical and biological studies.
  • Molecular formula C22H23N3O6S; molecular weight 457.50 g/mol.
  • Racemic isomer useful for stereochemistry and comparative assays.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Powder storage: -20°C (3 years) or 4°C (2 years); solvent stability varies.

Catalog No. 50-002-61093


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.