Learn More
Medchemexpress LLC (rac)-Exatecan Intermediate 1 | 102978-40-5 | 99.9% | 263.25 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY42487A100MG
Encompass_Preferred
(rac)-Exatecan Intermediate 1 is an isomer of Exatecan Intermediate 1 (HY-42487). Exatecan Intermediate 1 (compound 6) is an intermediate of Exatecan (HY-13631), a camptothecin-based anticancer agent.
  • Inhibits tumor growth by interfering with the proliferation and division of tumor cells by interacting with DNA.
  • Primarily used in research into a variety of cancers including ovarian, lung and breast cancer.
Catalog No. 50-004-05193