Learn More
Apexbio Technology LLC Rac GTPase fragment 1mg
SDP
Supplier:  Apexbio Technology LLC A10861
Encompass_Preferred
Rac GTPase fragment is a synthetic peptide representing a segment of the small signaling protein Rac a member of the Rho family of GTPases It is designed to serve as a molecular tool to investigate Rac-mediated modulation of actin dynamics and kinase activation processes essential for cytoskeletal rearrangement cell-cycle progression differentiation and cell adhesion Rac GTPase fragment exerts its biological activity by serving as a probe for Rac-driven signaling pathways Based on these pharmacological properties Rac GTPase fragment holds research potential in studying tumor biology particularly the development and progression of melanoma and non-small-cell lung carcinoma
Catalog No. 50-000-93669