Learn More
Medchemexpress LLC (Rac)-Hydnocarpin | 51419-48-8 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN7199
(Rac)-Hydnocarpin is a flavonoid isolated from *Hydnocarpus anthelminthica*. This compound exhibits moderate cytotoxic effects on cancer cells. It has a molecular formula of C25H20O9 and a molecular weight of 464.42.
- Isolated from *Hydnocarpus anthelminthica*.
- Exhibits moderate cytotoxic effects on cancer cells.
- Molecular formula: C25H20O9.
- Molecular weight: 464.42.
- Solid appearance.
- Off-white to yellow color.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.