Learn More
Medchemexpress LLC (Rac)-IBT6A | 1412418-47-3 | 1 ML

Supplier: Medchemexpress LLC HY1303610MM/1ML
(Rac)-IBT6A is a racemate of IBT6A, an impurity of Ibrutinib, commonly used in research and laboratory applications. This product is stable under recommended storage conditions and is not classified as hazardous, making it suitable for synthesis and various research purposes.
- Stable under recommended storage conditions.
- Research use only.
- Not classified as hazardous.
- Appearance: Powder.
- Molecular formula: C22H22N6O.
- Molecular weight: 386.45.
- Target: Btk.
- Pathway: Protein tyrosine kinase/RTK.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.