Learn More
Medchemexpress LLC (Rac)-IBT6A | 1412418-47-3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1303650MG
Encompass_Preferred
(Rac)-IBT6A is a racemate of IBT6A, which is an impurity of Ibrutinib. It can be utilized in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor.
  • Can be used in synthesis of Ibrutinib and Ibrutinib-based activity-based probes (ABPs)
  • Racemate form of IBT6A
  • Serves as an impurity of Ibrutinib
  • Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM
Catalog No. 50-003-40650