Learn More
Medchemexpress LLC HY-13036 5mg Medchemexpress, (Rac)-IBT6A CAS:1412418-47-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130365MG
Encompass_Preferred
Medchemexpress, HY-13036 5mg (Rac)-IBT6A CAS:1412418-47-3 (Rac)-IBT6A is a racemate of IBT6A. IBT6A is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1236